77339
Pertussis toxin from Bordetella pertussis
Synonym(s):
Histamine-sensitizing factor, IAP, Islet Activating Protein, PTX, Pertussigen
About This Item
storage temp.
−20°C
Gene Information
Bordetella pertussis Tohama I ... ptxA(2665068) , ptxB(2665069) , ptxC(2665408)
Looking for similar products? Visit Product Comparison Guide
Application
Biochem/physiol Actions
Physical form
Other Notes
replaced by
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service