Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77248

Supelco

Methyl cedryl ketone

reference material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26O
CAS Number:
Molecular Weight:
246.39
EC Number:
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

≥65.0% (GC)

refractive index

n20/D 1.516 (lit.)

bp

272 °C (lit.)

density

0.997 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

shipped in

wet ice

storage temp.

−20°C

SMILES string

C[C@@H]1CC[C@@H]2C(C)(C)[C@@H]3C[C@]12CC(C(C)=O)=C3C

InChI

1S/C17H26O/c1-10-6-7-15-16(4,5)14-9-17(10,15)8-13(11(14)2)12(3)18/h10,14-15H,6-9H2,1-5H3/t10-,14-,15-,17-/m1/s1

InChI key

YBUIAJZFOGJGLJ-GWBBYGMBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

>212.0 °F

flash_point_c

> 100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service