Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76744

Supelco

(R)-(+)-α-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol

derivatization grade (chiral), LiChropur, ≥99.0%

Synonym(s):

(R)-(+)-1-(Pentafluorophenyl)ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6F5)OH
CAS Number:
Molecular Weight:
212.12
Beilstein/REAXYS Number:
5268492
MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade (chiral)

Quality Level

assay

≥99.0% (sum of enantiomers, GC)
≥99.0%

form

fibers

optical activity

[α]20/D +7.0±0.5°, c = 1% in pentane

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

mp

40-43 °C
41-42 °C (lit.)

SMILES string

C[C@@H](O)c1c(F)c(F)c(F)c(F)c1F

InChI

1S/C8H5F5O/c1-2(14)3-4(9)6(11)8(13)7(12)5(3)10/h2,14H,1H3/t2-/m1/s1

InChI key

WYUNHWKTLDBPLE-UWTATZPHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(R)-(+)-α-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol is high quality, useful chiral derivatization reagent for all analytical applications in the chiral field. It is specially selected to meet the requirements for derivatization reagents for enantiomeric excess determinations

Other Notes

Chiral derivatizing agent for GC; derivatives can be detected in high sensitivity by electron capture detection, GC-ECD, or negative ion MS, NI/CI-MS

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.O. Meese
Liebigs Ann. Chem., 2004-2004 (1986)
Zhiping Li et al.
Experimental and therapeutic medicine, 14(6), 6119-6124 (2017-12-30)
The complement-activated product, complement component 5a (C5a), is a potent inflammatory peptide with a broad spectrum of functions. In vivo and in vitro studies have demonstrated that C5a serves an important role in inflammation; however, the role of C5a in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service