Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76530

Sigma-Aldrich

Pentadecanolide

puriss., ≥99.0% (GC)

Synonym(s):

1-Oxa-2-cyclohexadecanone, 15-Hydroxypentadecanoic acid lactone, 15-Pentadecanolide, Pentalide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28O2
CAS Number:
Molecular Weight:
240.38
Beilstein/REAXYS Number:
143710
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

puriss.

Quality Level

assay

≥99.0% (GC)

form

crystals

bp

137 °C/2 mmHg (lit.)

mp

34-36 °C
34-38 °C (lit.)

density

0.918 g/mL at 25 °C (lit.)

SMILES string

O=C1CCCCCCCCCCCCCCO1

InChI

1S/C15H28O2/c16-15-13-11-9-7-5-3-1-2-4-6-8-10-12-14-17-15/h1-14H2

InChI key

FKUPPRZPSYCDRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

320.0 °F - closed cup

flash_point_c

160 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service