76442
Pentaethylene glycol monotetradecyl ether
≥99.0% (TLC)
Synonym(s):
C14E5, Polyoxyethylene (5) myristyl ether, Tetradecyl pentaethylene glycol ether, Tetradecylpentaglycol
About This Item
description
non-ionic
assay
≥99.0% (TLC)
form
liquid
mol wt
434.65 g/mol
mp
32-33 °C
cation traces
Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cr: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg
SMILES string
CCCCCCCCCCCCCCOCCOCCOCCOCCOCCO
InChI
1S/C24H50O6/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-26-17-19-28-21-23-30-24-22-29-20-18-27-16-14-25/h25H,2-24H2,1H3
InChI key
KXGOCXPKKMVOSB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service