Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

7620031500BC

Millipore

Gemini Reagent Rack Adapters

BioControl Systems, for use with Gemini®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.21
Product 7620031500BC is not currently sold in your country. Contact Technical Service

packaging

pkg of 6 units

manufacturer/tradename

BioControl Systems
Gemini®

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

application(s)

food and beverages

compatibility

for use with Gemini®

storage temp.

room temp

Legal Information

Gemini is a registered trademark of Stratec

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service