76034
HypoGel® 200 NH2
extent of labeling: ~0.9 mmol/g loading
Synonym(s):
O-(2-Aminoethyl)tetra(oxyethylene) polymer-bound
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.21
form
crystals
extent of labeling
~0.9 mmol/g loading
particle size
110-150 μm
storage temp.
2-8°C
Legal Information
HypoGel is a registered trademark of Rapp Polymere GmbH
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service