Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75806

Millipore

Oxford Listeria Selective Supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

Application

An antimicrobial supplement recommended for the selective isolation of Listeria monocytogenes from specimens containing a mixed bacterial flora.

Packaging

Set with 10 vials.

Components

Composition: Fosfomycin 5 mg, Cefotetan 1 mg, Colistin sulfate 10 mg, Cycloheximide 200 mg, Acriflavine 2.5 mg

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 2 - Muta. 2 - Repr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service