Skip to Content
MilliporeSigma

75752

Dimethyl oxalate

purum, ≥99.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3OCOCOOCH3
CAS Number:
Molecular Weight:
118.09
EC Number:
209-053-6
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1071744
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥99.0% (GC)

refractive index

n20/D 1.39 (lit.)

bp

163.5 °C (lit.)

mp

50-54 °C (lit.)

density

1.148 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(=O)OC

InChI

1S/C4H6O4/c1-7-3(5)4(6)8-2/h1-2H3

InChI key

LOMVENUNSWAXEN-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under Dimethyl oxalate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
11317-1KG04061838089236
806022000504022536388854
806022252104061839611597
806022500004022536912912
471356-5ML04061837548956
471356-100ML04061837548925
471356-500ML04061837548932
471356-5L04061837548949
471356-25ML04061832359939
471348-100ML04061832359892
M4141-10X2ML04061837841088
806022025004022536388861
806022250104061839611580
806022052104061839611573
806022902504022536781242
806022100004022536388878
806022999904022536388885
471348-1L04061832359908
471348-500ML04061832359915
471356-20L04022536026213