Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75571

Sigma-Aldrich

Trimethyl orthobutyrate

purum, ≥97.0% (GC)

Synonym(s):

1,1,1-Trimethoxybutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2C(OCH3)3
CAS Number:
Molecular Weight:
148.20
Beilstein/REAXYS Number:
1737319
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.404 (lit.)
n20/D 1.404

bp

145-147 °C (lit.)

density

0.926 g/mL at 25 °C (lit.)

SMILES string

CCCC(OC)(OC)OC

InChI

1S/C7H16O3/c1-5-6-7(8-2,9-3)10-4/h5-6H2,1-4H3

InChI key

JAFMOTJMRSZOJE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethyl orthobutyrate can be used for:
  • Acylation at OH-9 of N-acetylneuraminic acid and N-glycoloylneuraminic acid to give the corresponding 9-O-acylated derivatives.[1]
  • Conversion of 2-amino-N-(1-H-benzimidazol-2-yl)benzamide to 2-propyl-3-benzimidazolyl-4(3H)-quinazolinone.[2]
  • Preparation of 2-propyl substituted benzimidazoles from 1,2-benzenediamines.[3]

It can also be used in the total synthesis of dl-camptothecin[4], Olmesartan Medoxomil[5], DE and CDE ring analogs of camptothecin[6], and homologs of 1,25-dihydroxyvitamin D3.[7]

Other Notes

Reagent for preparing cyclic 17,21-methyl orthobutyrates of corticoids[8][9]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

122.0 °F - closed cup

flash_point_c

50 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of new PPARγ agonists: Benzimidazole derivatives-importance of positions 5 and 6, and computational studies on the binding mode.
Goebel M, et al.
Bioorganic & Medicinal Chemistry, 18(16), 5885-5895 (2010)
Total synthesis of dl-camptothecin.
Tang C S, et al.
Journal of the American Chemical Society, 97(1), 159-167 (1975)
Unique rearrangement of ergocalciferol side chain in vitro: production of a biologically highly active homologue of 1, 25-dihydroxyvitamin D3.
Tanaka Y, et al.
Biochemistry, 25(19), 5512-5518 (1986)
Efficient synthesis of olmesartan medoxomil, an antihypertensive drug.
Babu K S, et al.
Synthetic Communications, 39(2), 291-298 (2008)
Synthesis of some DE and CDE ring analogs of camptothecin.
Plattner J J, et al.
Journal of the American Chemical Society, 94(24), 8613-8615 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service