Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

75142

Supelco

Hypophyllanthin

analytical standard

Synonym(s):

NSC 619044

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30O7
CAS Number:
Molecular Weight:
430.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24
Product 75142 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

COC1=CC([C@@H]2C3=C4C(OCO4)=C(OC)C=C3C[C@H](COC)[C@H]2COC)=CC=C1OC

InChI

1S/C24H30O7/c1-25-11-16-8-15-10-20(29-5)23-24(31-13-30-23)22(15)21(17(16)12-26-2)14-6-7-18(27-3)19(9-14)28-4/h6-7,9-10,16-17,21H,8,11-13H2,1-5H3/t16-,17-,21+/m1/s1

InChI key

LBJCUHLNHSKZBW-LZJOCLMNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akilandeshwari Alagan et al.
Frontiers in pharmacology, 10, 632-632 (2019-06-25)
Background:Phyllanthus amarus (PA) is widely studied for its hepatoprotective properties but has recently received increasing attention due to its diverse anti-inflammatory effects. However, the effects of PA in modulating immune responses in the central nervous system leading to protection against
In Vitro Chondroprotective Potential of Extracts Obtained from Various Phyllantus Species.
Kittisak Buddhachat et al.
Planta medica, 83(1-02), 87-96 (2016-06-25)
Cherdsak Boonyong et al.
The Journal of pharmacy and pharmacology, 72(4), 583-591 (2020-01-11)
Indomethacin (INDO) and diclofenac (DIC) can induce intestinal cell death through induction of oxidative stress-mediated ER stress and mitochondrial dysfunction. This study investigated the cytoprotective potential of 11 polyphenols, namely caffeic acid (CAF), curcumin (CUR), epigallocatechin gallate (EGCG), gallic acid
V Suresh et al.
Journal of ethnopharmacology, 156, 309-315 (2014-09-16)
Phyllanthusrheedei Wight is a plant used by Muthuvan tribes of Kerala for treating liver related diseases. The different extracts of Phyllanthus rheedei were analysed on cell lines were viz, PLC/PRF, Hep3B, FLCII10 and HepG2215 for its anti-HBV property. The analysis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service