Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75002

Sigma-Aldrich

4-Octylbenzoic acid

puriss., liquid crystal, ≥99.0% (T)

Synonym(s):

4-n -Octylbenzoic acid, p -Octylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7C6H4CO2H
CAS Number:
Molecular Weight:
234.33
Beilstein/REAXYS Number:
1954607
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

grade

puriss.

assay

≥99.0% (T)

form

liquid crystal

mp

97-98 °C (lit.)
97-98 °C

SMILES string

CCCCCCCCc1ccc(cc1)C(O)=O

InChI

1S/C15H22O2/c1-2-3-4-5-6-7-8-13-9-11-14(12-10-13)15(16)17/h9-12H,2-8H2,1H3,(H,16,17)

InChI key

ZQLDNJKHLQOJGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service