75000
tert-Octylamine
technical, ≥95% (GC)
Synonym(s):
1,1,3,3-Tetramethylbutylamine, 2-Amino-2,4,4-trimethylpentane
About This Item
Recommended Products
vapor density
4.46 (vs air)
vapor pressure
10 mmHg ( 25 °C)
grade
technical
assay
≥95% (GC)
form
liquid
refractive index
n20/D 1.424 (lit.)
n20/D 1.425
bp
137-143 °C (lit.)
density
0.805 g/mL at 25 °C (lit.)
SMILES string
C\C(C)=N/O
CC(C)(C)CC(C)(C)N
InChI
1S/C8H19N/c1-7(2,3)6-8(4,5)9/h6,9H2,1-5H3
InChI key
QIJIUJYANDSEKG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
replaced by
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1C
Storage Class
3 - Flammable liquids
wgk_germany
WGK 2
flash_point_f
91.4 °F - closed cup
flash_point_c
33 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service