Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

74907

Supelco

Ferimzone

PESTANAL®, analytical standard

Synonym(s):

(Z)-2′-Methylacetophenone 4,6-dimethylpyrimidin-2-ylhydrazone, 4,6-Dimethyl-2(1H)-pyrimidinone (2Z)-[1-(2-methylphenyl)ethylidene]hydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4
CAS Number:
Molecular Weight:
254.33
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

composition

carbon content, 70.84%
hydrogen content, 7.13%
nitrogen content, 22.03%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

172-177 °C

application(s)

agriculture
environmental

format

neat

InChI

1S/C15H18N4/c1-10-7-5-6-8-14(10)13(4)18-19-15-16-11(2)9-12(3)17-15/h5-9H,1-4H3,(H,16,17,19)/b18-13-

InChI key

GOWLARCWZRESHU-AQTBWJFISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ferimzone is a novel, systemic fungicide,[1] widely used in rice fields for the control of rice blast and leaf spot.[2]

Application

Ferimzone may be used as an analytical reference standard for the quantification of the analyte in apples,[3] lettuce samples,[3] fruit and root vegetables[4] using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 2

target_organs

Nervous system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Pesticide Encyclopedia (2014)
Monitoring of pesticide residues and risk assessment for fruit vegetables and root vegetables of environment-friendly certified and general agricultural products
Ahn W-J
Korean Journal of Environmental Agriculture, 31(2), 164-169 (2012)
Rice Blast: Interaction with Rice and Control: Proceedings of the 3rd International Rice Blast Conference (2004)
Multiresidue analysis of 240 pesticides in apple and lettuce by QuEChERS sample preparation and HPLC-MS/MS analysis
Hye-Young K, et al.
The Korean Journal of Pesticide Science, 15(4), 417-433 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service