Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C38H47NO6
CAS Number:
Molecular Weight:
613.78
UNSPSC Code:
12352200
MDL number:
assay
≥98.0% (HPCE)
solubility
DMSO: soluble, acetonitrile: soluble
fluorescence
λex 497 nm; λem 519 nm in methanol
suitability
suitable for fluorescence
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCCC(=O)Nc1ccc(c(c1)C(O)=O)C2=C3C=CC(=O)C=C3Oc4cc(O)ccc24
InChI
1S/C38H47NO6/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-36(42)39-27-18-21-30(33(24-27)38(43)44)37-31-22-19-28(40)25-34(31)45-35-26-29(41)20-23-32(35)37/h18-26,40H,2-17H2,1H3,(H,39,42)(H,43,44)
InChI key
XKFGZSAKZAPLGI-UHFFFAOYSA-N
Other Notes
Lipophilic probe for studying membrane lipid behaviour by fluorescence recovery after photobleaching
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
D A McKay et al.
Journal of cell science, 100 ( Pt 3), 473-479 (1991-11-01)
Differences in membrane composition between the anterior and posterior poles of human blood neutrophils on exposure to chemoattractant were studied using a laser microscope, and the effects of attractant concentration and time on the ability of neutrophils to polarize were