Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74549

Supelco

Gluconate Standard for IC

TraceCERT®, 1000 mg/L gluconate in water (nominal concentration)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1000 mg/L gluconate in water (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L glycolate in water, prepared with glycolic acid and watera; filtered through a 0.2 μm membrane filter and stabilized with sodium azide

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service