Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74348

Supelco

4-Hydroxybenzyl alcohol

analytical standard

Synonym(s):

NSC 227926

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CH2OH
CAS Number:
Molecular Weight:
124.14
Beilstein/REAXYS Number:
1858967
EC Number:
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

mp

114-122 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OCc1ccc(O)cc1

InChI

1S/C7H8O2/c8-5-6-1-3-7(9)4-2-6/h1-4,8-9H,5H2

InChI key

BVJSUAQZOZWCKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service