Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74203

Millipore

Gram+ / Gram- Differentiation Enzyme Confirmation Test

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129

shelf life

limited shelf life, expiry date on the label

Quality Level

application(s)

food and beverages
microbiology

Application

This tests utilizes a substrate which, when hydrolyzed by a specific enzyme (during peptide hydrolysis), produces together with a developer a blue color.

Components

1 Bottle of Reagent A (Buffer)
1 Bottle of Reagent B (Developer)
12 Test Cards with 4 spots

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service