Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74198

Supelco

trans,trans-2,4-Hexadienal

analytical standard

Synonym(s):

Sorbaldehyde, Sorbic aldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCHO
CAS Number:
Molecular Weight:
96.13
Beilstein/REAXYS Number:
1698401
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.541 (lit.)
n20/D 1.541

bp

69 °C/20 mmHg (lit.)

density

0.895 g/mL at 20 °C
0.871 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

[H]C(=O)\C=C\C=C\C

InChI

1S/C6H8O/c1-2-3-4-5-6-7/h2-6H,1H3/b3-2+,5-4+

InChI key

BATOPAZDIZEVQF-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service