Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74182

Supelco

2,5-Dimethylthiophene

analytical standard

Synonym(s):

NSC 60689

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8S
CAS Number:
Molecular Weight:
112.19
Beilstein/REAXYS Number:
106450
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

refractive index

n20/D 1.510-1.514
n20/D 1.512 (lit.)

bp

134 °C/740 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1ccc(C)s1

InChI

1S/C6H8S/c1-5-3-4-6(2)7-5/h3-4H,1-2H3

InChI key

GWQOOADXMVQEFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

75.2 °F - closed cup

flash_point_c

24 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service