Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74132

Supelco

Isofucoxanthinol

analytical standard

Synonym(s):

6,6′,7,7′-Tetradehydro-4′,5,5′,6,7,8-hexahydro-3,3′,5,5′-tetrahydroxy-8-oxo-β,​ε-carotene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H56O5
CAS Number:
Molecular Weight:
616.87
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

General description

Isofucoxanthinol belongs to the class of carotenoids. It is an allenic pigment of fucoxanthin, a common algal carotenoid.

Application

Isofucoxanthinol may be used as an analytical standard for the determination of the analyte in sediment traps and edible brown seaweeds by high-performance liquid chromatography (HPLC) technique.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suppressive effects of amarouciaxanthin A on 3T3-L1 adipocyte differentiation through down-regulation of PPAR\gamma and C/EBPalpha mRNA expression
Yim MJ, et al.
Journal of Agricultural and Food Chemistry, 59(5), 1646-1652 (2011)
Neoxanthin and fucoxanthinol in Fucus vesiculosus
Nitsche H
Biochimica et biophysica acta. General subjects, 338(2), 572-576 (1974)
Fucoxanthin and related pigments
Bonnett R, et al.
Chemical Communications (Cambridge, England), 119(15), 515-516 (1966)
Carotenoid transformations in coastal marine waters
Repeta DJ and Gagosian RB
Nature, 295(5844), 51-54 (1982)
Fucoxanthin metabolites in egg yolks of laying hens
Strand A, et al.
Comparative Biochemistry and Physiology. Part A, Molecular & Integrative Physiology, 119(4), 963-974 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service