Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74131

Supelco

2-Imidazolidinethione-4,4,5,5-d4

PESTANAL®, analytical standard

Synonym(s):

2-Thioxoimidazolidine-4,4,5,5-d4, N,N′-Ethylene-d4-thiourea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3D4H2N2S
CAS Number:
Molecular Weight:
106.18
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥95 atom % D

product line

PESTANAL®

assay

≥97.5% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

InChI

1S/C3H6N2S/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)/i1D2,2D2

InChI key

PDQAZBWRQCGBEV-LNLMKGTHSA-N

General description

2-Imidazolidinethione-4,4,5,5-d4 is an isotope labeled analog of the fungicide metabolite 2-imidazolidinethione, wherein the imidazoline ring C-4 and C-5 protons are replaced by deuterium.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis, potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rodenticides
Szycher's Handbook of Polyurethanes, 2153-2217 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service