74038
2-Undecanone
analytical standard
Synonym(s):
Methyl nonyl ketone
About This Item
grade
analytical standard
Quality Level
vapor density
5.9 (vs air)
vapor pressure
<1 mmHg ( 20 °C)
assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.428-1.432
n20/D 1.43 (lit.)
bp
231-232 °C (lit.)
mp
11-13 °C (lit.)
density
0.825 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
food and beverages
personal care
format
neat
SMILES string
CCCCCCCCCC(C)=O
InChI
1S/C11H22O/c1-3-4-5-6-7-8-9-10-11(2)12/h3-10H2,1-2H3
InChI key
KYWIYKKSMDLRDC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class
10 - Combustible liquids
wgk_germany
WGK 2
flash_point_f
192.2 °F - closed cup
flash_point_c
89 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service