Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74034

Supelco

2-Tridecanone

analytical standard

Synonym(s):

Methyl undecyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COCH3
CAS Number:
Molecular Weight:
198.34
Beilstein/REAXYS Number:
1757402
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.435 (lit.)

bp

134 °C/10 mmHg (lit.)
263 °C (lit.)

mp

24-27 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCC(C)=O

InChI

1S/C13H26O/c1-3-4-5-6-7-8-9-10-11-12-13(2)14/h3-12H2,1-2H3

InChI key

CYIFVRUOHKNECG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Tridecanone is an organic compound belonging to the class of ketones. It is a food aroma compound, widely used in the flavor and fragrance industry.

Application

2-Tridecanone may be used as an analytical reference standard for the quantification of the analyte in the following:        
  • Acerola fruit using GC and GC-MS techniques.        
  • Rosa hybrida using gas-chromatography coupled to mass spectrometry (GC-MS) technique.       
  • Ginger oil using capillary GC and GC-MS techniques.

pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

224.6 °F - closed cup

flash_point_c

107 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natural and Artificial Flavoring Agents and Food Dyes
Handbook of Food Bioengineering, 7 (2017)
Bérengère de Laval et al.
Cell stem cell, 26(5), 657-674 (2020-03-15)
Hematopoietic stem cells (HSCs) maintain life-long production of immune cells and can directly respond to infection, but sustained effects on the immune response remain unclear. We show that acute immune stimulation with lipopolysaccharide (LPS) induced only transient changes in HSC
Gas chromatographic analysis of volatile components of ginger oil (Zingiber officinale Roscoe) extracted with liquid carbon dioxide
Chen C-C, et al.
Journal of Agricultural and Food Chemistry, 36(2), 322-328 (1988)
Fundamentals of Food Biotechnology, 2 (2009)
Determination of floral fragrances of Rosa hybrida using solid-phase trapping-solvent extraction and gas chromatography?mass spectrometry
Kim JH, et al.
Journal of Chromatography A, 902(2), 389-404 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service