Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74009

Supelco

1-Decanoyl-rac-glycerol

analytical standard

Synonym(s):

Monocaprin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H26O4
CAS Number:
Molecular Weight:
246.34
EC Number:
MDL number:
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

assay

≥99.0% (GC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

functional group

ketone

Storage temp.

−20°C

SMILES string

O=C(OCC(O)CO)CCCCCCCCC

InChI

1S/C13H26O4/c1-2-3-4-5-6-7-8-9-13(16)17-11-12(15)10-14/h12,14-15H,2-11H2,1H3

Inchi Key

LKUNXBRZDFMZOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Decanoyl-rac-glycerol is a food additive, widely used as a functional emulsifier in food industry. It exhibits a broad-spectrum of activity against a variety of food-borne bacteria, fungi and viruses.[1]

Application

1-Decanoyl-rac-glycerol may be used as an analytical standard in determining the purity of the analyte synthesized from camphor tree seed kernel oil and glycerol using the lipase from porcine pancreas as a catalyst by gas chromatography technique.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antifungal activity and mechanism of monocaprin against food spoilage fungi
Ma M, et al.
Food Control, 84, 561-568 (2018)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service