Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

74005

Supelco

DL-3-Ureidoisobutyric acid

≥99.0% (HPLC)

Synonym(s):

β-UBA, β-Ureidoisobutyric acid, 3-(Carbamoylamino)-2-methylpropanoic acid, 3-Ureido-2-methylpropanoic acid, 3-[(Aminocarbonyl)amino]-2-methylpropanoic acid, N-Carbamoyl-2-methyl-β-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O3
CAS Number:
Molecular Weight:
146.14
Beilstein/REAXYS Number:
1768736
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥99.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

neat

Storage temp.

2-8°C

SMILES string

OC(C(C)CNC(N)=O)=O

InChI

1S/C5H10N2O3/c1-3(4(8)9)2-7-5(6)10/h3H,2H2,1H3,(H,8,9)(H3,6,7,10)

Inchi Key

PHENTZNALBMCQD-UHFFFAOYSA-N

Biochem/physiol Actions

3-Ureidoisobutyrate is increased in the urine of patients with beta-ureidopropionase deficiency and can be used to predict patient′s individual phenotypes of enzyme deficiencies in pyrimidine metabolism when associated with a risk for severe toxicity against the antineoplastic agent 5-fluorouracil.

Pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morimasa Ohse et al.
Journal of mass spectrometry : JMS, 37(9), 954-962 (2002-09-25)
Dihydropyrimidine dehydrogenase (DHPDase), dihydropyrimidinase (DHPase) and beta-ureidopropionase (betaUPase) are the enzymes that catalyze the first, second, and third steps of the degradation of pyrimidines, respectively. beta-Ureidopropionate (betaUP) and beta-ureidoisobutyrate (betaUIB) are increased in the urine of patients with betaUPase deficiency.
Ute Hofmann et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 791(1-2), 371-380 (2003-06-12)
Enzyme deficiencies in pyrimidine metabolism are associated with a risk for severe toxicity against the antineoplastic agent 5-fluorouracil. To assess whether urinary levels of pyrimidines and their metabolites can be used for predicting patients' individual phenotype, a new gas chromatographic-tandem
André B P van Kuilenburg et al.
Human molecular genetics, 13(22), 2793-2801 (2004-09-24)
beta-Ureidopropionase deficiency is an inborn error of the pyrimidine degradation pathway, affecting the cleavage of N-carbamyl-beta-alanine and N-carbamyl-beta-aminoisobutyric acid. In this study, we report the elucidation of the genetic basis underlying a beta-ureidopropionase deficiency in four patients presenting with neurological

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service