Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73929

Supelco

Sodium ionophore VIII

Selectophore, function tested

Synonym(s):

Bis[(12-crown-4)methyl] 2,2-didodecylmalonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H84O12
CAS Number:
Molecular Weight:
817.14
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

product line

Selectophore

Quality Level

quality

function tested

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCC(CCCCCCCCCCCC)(C(=O)OCC1COCCOCCOCCO1)C(=O)OCC2COCCOCCOCCO2

InChI

1S/C45H84O12/c1-3-5-7-9-11-13-15-17-19-21-23-45(24-22-20-18-16-14-12-10-8-6-4-2,43(46)56-39-41-37-52-31-29-48-25-27-50-33-35-54-41)44(47)57-40-42-38-53-32-30-49-26-28-51-34-36-55-42/h41-42H,3-40H2,1-2H3

InChI key

PQDPQSWXOQEQHB-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Other Notes

Highly lipophilic and selective carrier for sodium selective electrodes[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of α-Substitutents on Ion Selectivity of Bis(12-crown-4-methyl) Malonates as Neutral Carriers for Sodium Ion-Selective Electrodes
Kimura, K., et al.
Analytical Sciences, 12, 67-67 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service