Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73829

Supelco

(±)-Ureidoglycolic acid sodium salt

analytical standard

Synonym(s):

2-Hydroxyhydantoic acid sodium salt, Sodium ureidoglycolate, [(Aminocarbonyl)amino]hydroxyacetic acid monosodium salt

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C3H5N2NaO4
CAS Number:
Molecular Weight:
156.07
UNSPSC Code:
12352200
NACRES:
NB.21

grade

analytical standard

Quality Level

assay

≥95.0% (TLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

NC(NC(O)C(O[Na])=O)=O

InChI

1S/C3H6N2O4/c4-3(9)5-1(6)2(7)8/h1,6H,(H,7,8)(H3,4,5,9)/t1-/m1/s1

InChI key

NWZYYCVIOKVTII-PVQJCKRUSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service