Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73638

Supelco

Cation Calibration Standard for MARGA (Metrohm Air Monitoring Instrument)

TraceCERT®, 6 cations, 0.25 mg/L in nitric acid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

TraceCERT®

concentration

0.25 mg/L in nitric acid

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST or BAM.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Analysis Note

0.25 mg/L each: Li+, Na+, K+, Mg2+, Ca2+ and NH4+, in <0.1% nitric acid

Other Notes

For a complete product listing of our TraceCERT range of CRMs for ICP and AAS, technical information and example certificates please visit our inorganic TraceCERT website

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service