Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73536

Supelco

Acetovanillone

analytical standard

Synonym(s):

4′-Hydroxy-3′-methoxyacetophenone, Acetovanillone, Apocynin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
Beilstein/REAXYS Number:
637373
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

263-265 °C/17 mmHg (lit.)

mp

112-115 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

COc1cc(ccc1O)C(C)=O

InChI

1S/C9H10O3/c1-6(10)7-3-4-8(11)9(5-7)12-2/h3-5,11H,1-2H3

InChI key

DFYRUELUNQRZTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetovanillone is an aromatic plant ketone, which is a selective inhibitor of NADPH oxidase.[1] It is mainly secreted in the urine of mammals as the corresponding glucuronide conjugate.[2]

Application

Acetovanillone may be used as a reference standard for the determination of the analyte in Vanilla planifolia using reversed-phase high-performance liquid chromatography (RP-HPLC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Mammalian Metabolism of Plant Compounds (1991) (2017)
The NADPH oxidase inhibitor apocynin (acetovanillone) induces oxidative stress.
Riganti C, et al.
Toxicology and Applied Pharmacology, 212(3), 179-187 (2006)
Development and validation of an RP?HPLC method for quantitative determination of vanillin and related phenolic compounds in Vanilla planifolia.
Sinha KA, et al.
Journal of Separation Science, 30(1), 15-20 (2007)
Hye Kyoung Sung et al.
Scientific reports, 9(1), 4668-4668 (2019-03-16)
Iron overload is associated with various pathological changes which contribute to heart failure. Here, we examined mechanisms via which iron alters cardiomyocyte insulin sensitivity. Treatment of primary adult and neonatal cardiomyocytes as well as H9c2 cells with iron decreased insulin

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service