Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73535

Supelco

2-Acetylthiophene

analytical standard

Synonym(s):

Methyl-2-thienyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6OS
CAS Number:
Molecular Weight:
126.18
Beilstein/REAXYS Number:
107910
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

refractive index

n20/D 1.565 (lit.)
n20/D 1.565-1.568

bp

214 °C (lit.)

mp

10-11 °C (lit.)

density

1.168 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)c1cccs1

InChI

1S/C6H6OS/c1-5(7)6-3-2-4-8-6/h2-4H,1H3

InChI key

WYJOVVXUZNRJQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

206.6 °F

flash_point_c

97 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mostafa M Ghorab et al.
Acta pharmaceutica (Zagreb, Croatia), 64(4), 419-431 (2014-12-23)
A novel series of thiophenes having biologically active sulfonamide 2-11, 3-methylisoxazole 12, 4-methoxybenzo[d] thiazole 13, quinoline 14, 15, benzoylphenylamino 16, and anthracene-9,10-dione 17 moieties were prepared. Structures of the newly synthesized compounds were established by elemental analysis and spectral data.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service