Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73510

Sigma-Aldrich

1-Nitropentane

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4NO2
CAS Number:
Molecular Weight:
117.15
Beilstein/REAXYS Number:
506508
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

purum

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.417 (lit.)
n20/D 1.417

bp

75-76 °C/23 mmHg (lit.)

density

0.952 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCC[N+]([O-])=O

InChI

1S/C5H11NO2/c1-2-3-4-5-6(7)8/h2-5H2,1H3

InChI key

BVALZCVRLDMXOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

141.8 °F - closed cup

flash_point_c

61 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service