Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73154

Supelco

2,2-Di(2-tetrahydrofuryl)propane, mixture of isomers

analytical standard

Synonym(s):

2,2′-Isopropylidenebis(tetrahydrofuran), 2,2-Bis(2-oxolanyl)propane, 2,2-Ditetrahydrofurylpropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28
Beilstein/REAXYS Number:
6855883
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24
Product 73154 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.468-1.472

format

neat

InChI

1S/C11H20O2/c1-11(2,9-5-3-7-12-9)10-6-4-8-13-10/h9-10H,3-8H2,1-2H3/t9-,10-/m0/s1

InChI key

FZLHAQMQWDDWFI-UWVGGRQHSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

supp_hazards

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

221.0 °F

flash_point_c

105 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service