73104
O-Phosphorylethanolamine
certified reference material, TraceCERT®
Synonym(s):
2-Aminoethyl dihydrogen phosphate, O-Phosphocolamine, O-Phosphoethanolamine, Colaminephosphoric acid
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
format
neat
storage temp.
−20°C
SMILES string
NCCOP(O)(O)=O
InChI
1S/C2H8NO4P/c3-1-2-7-8(4,5)6/h1-3H2,(H2,4,5,6)
InChI key
SUHOOTKUPISOBE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.
Recommended products
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service