Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73095BC

GDS HT Upgrade Package

BioControl, For use with GDS HighThroughput (small 0.1 μL amplification tubes)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

manufacturer/tradename

BioControl Systems 73095

storage condition

room temperature

compatibility

BioControl GDS

storage temp.

room temp

Features and Benefits

For use with GDS 576 kits

Components

  • Rotor (72-well, Rotor-Gene Q)
  • Locking ring (72-well, Rotor-Gene Q)
  • Amp tube capping tool, 72 well
  • Aluminum cooling block, 72 well
  • Variable spacing amp tube holder, 72 well
  • Variable spacing amp tube holder lid, 72 well

Testing Method

PCR

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service