Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73002

Supelco

Butyl stearate

analytical standard

Synonym(s):

Stearic acid butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)16COO(CH2)3CH3
CAS Number:
Molecular Weight:
340.58
Beilstein/REAXYS Number:
1792866
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

density

0.861 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCCCC

InChI

1S/C22H44O2/c1-3-5-7-8-9-10-11-12-13-14-15-16-17-18-19-20-22(23)24-21-6-4-2/h3-21H2,1-2H3

InChI key

ULBTUVJTXULMLP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butyl stearate is a fatty acid ester, which has application in cosmetics, personal care products, and as an emollient in food industries.[1][2][3]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Butyl stearate may be used as an analytical standard for the determination of the analyte in cosmetic products[4][5] and fruit-based baby foods[1] by various chromatography techniques. It may be used as an internal standard for the determination of fungicides in fruit juice and wine samples by capillary gas chromatography[6] and for the analysis of lysinoalanine in food proteins by gas-liquid chromatography (GLC).[7]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

320.0 °F - closed cup

flash_point_c

160 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Proceedings of the World Conference on Palm and Coconut Oils for the 21st Century: Sources, Processing, Applications, and Competition
Proceedings of the World Conference on Palm and Coconut Oils for the 21st Century (1999)
Trace analysis of fungicides in grape juice and wine using glass capillary gas chromatography
Spitzer T and Nickless G
Journal of High Resolution Chromatography, 4(4), 151-155 (1981)
Gas-liquid chromatography of some fatty acid esters and alcohols in lipsticks
Gross FC
Journal of the Association of Official Analytical Chemists, 49(6), 1196-1200 (1966)
Studies on the gas chromatographic analysis of lysinoalanine in alkali-treated food proteins
Hasegawa K and Okamoto N
Agricultural and Biological Chemistry, 44(3), 649-655 (1980)
A environmentally friendly reversed-phase liquid chromatography method for phthalates determination in nail cosmetics
De Orsi, et al.
Analytica Chimica Acta, 555(2), 238-241 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service