Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72780

Sigma-Aldrich

2-Nitrobenzaldehyde

for the determination of methyl ketones, ≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CHO
CAS Number:
Molecular Weight:
151.12
Beilstein/REAXYS Number:
742624
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥99.0%

form

crystals

quality

for the determination of methyl ketones

ign. residue

≤0.1% (as SO4)

bp

153 °C/23 mmHg (lit.)

mp

42-44 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤200 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤25 mg/kg
Fe: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤30 mg/kg
Zn: ≤20 mg/kg

SMILES string

[O-][N+](=O)c1ccccc1C=O

InChI

1S/C7H5NO3/c9-5-6-3-1-2-4-7(6)8(10)11/h1-5H

InChI key

CMWKITSNTDAEDT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An-Hu Li et al.
Organic & biomolecular chemistry, 5(1), 61-64 (2006-12-14)
A highly effective one-pot Friedländer quinoline synthesis using inexpensive reagents has been developed. o-Nitroarylcarbaldehydes were reduced to o-aminoarylcarbaldehydes with iron in the presence of catalytic HCl (aq.) and subsequently condensed in situ with aldehydes or ketones to form mono- or
Shibo Cheng et al.
Physical chemistry chemical physics : PCCP, 12(31), 9067-9074 (2010-06-08)
The fluorescence and phosphorescence emission studies have been performed to understand the photochemistry of o-nitrobenzaldehyde in solution upon photoexcitation to the first excited singlet state (S(1)). The presence of an energy barrier in the S(1)-state hydrogen transfer potential energy surface
The photochemistry of o-nitrobenzaldehyde as seen by femtosecond vibrational spectroscopy.
Stefan Laimgruber et al.
Angewandte Chemie (International ed. in English), 44(48), 7901-7904 (2005-11-16)
J M Allen et al.
Journal of pharmaceutical and biomedical analysis, 24(2), 167-178 (2000-12-29)
We report the development of a new 'photochemical titration' actinometric method for measurement of UV-B (290-320 nm) and UV-A (320-400 nm) light dose during drug photostability testing. It is based upon photolysis of aqueous 2-nitrobenzaldehyde solution, a well-characterized reaction that
Gregory O Noonan et al.
Journal of agricultural and food chemistry, 53(12), 4680-4685 (2005-06-09)
Recently, semicarbazide has been found in food in jars sealed with cap liners that were manufactured using azodicarbonamide as a blowing agent. These reports raised the concern that the use of azodicarbonamide-an approved dough conditioner-may result in semicarbazide residues in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service