Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72743

Supelco

S-(2-Succinyl)-L-cysteine

analytical standard

Synonym(s):

2-[[(2R)-2-Amino-2-carboxyethyl]thio]butanedioic acid, S-(1,2-Dicarboxyethyl)-L-cysteine, S-(2-Succino)-L-cysteine, [[(2R)-2-Amino-2-carboxyethyl]thio]succinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11NO6S
CAS Number:
Molecular Weight:
237.23
Beilstein/REAXYS Number:
1729068
UNSPSC Code:
12352200

grade

analytical standard

Quality Level

assay

≥95.0% (TLC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

technique(s)

LC/MS: suitable

color

white to off-white

format

neat

Storage temp.

2-8°C

SMILES string

O=C([C@@](C([H])([H])S[C@@](C(O[H])=O)([H])C([H])([H])C(O[H])=O)([H])N([H])[H])O[H]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the stereochemistry of S-(2-Succinyl)-L-cysteine (P/N: 72743) and S-(1,2-Dicarboxyethyl)glutathione (P/N: 91938)? Specifically, does the "S" represent sulfur or chiral configuration?

    1 answer
    1. The “S” & “R” in the nomenclature of these molecules solely represent information regarding the stereochemistry, and this information is not related to the sulfur atom present in this molecule.
      72743 has two chiral centers: one is carbon 2 on L-cysteine. The other chiral center should refer to the carbon on 2-succinyl group which connects to the sulfur. 91938 has one chiral center, which is the same one on 2-succinyl group.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service