Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

72702

Supelco

3,5-Dinitrobenzoyl chloride

derivatization grade (HPLC), LiChropur

Synonym(s):

DNBC, NSC 2697

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)2C6H3COCl
CAS Number:
Molecular Weight:
230.56
Beilstein/REAXYS Number:
990249
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade (HPLC)

Quality Level

vapor density

7.6 (vs air)

assay

≥98.0% (GC)
98.0-102.0% (AT)

form

crystals

quality

LiChropur

technique(s)

HPLC: suitable

bp

196 °C/11 mmHg (lit.)

mp

68-69 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(cc(c1)[N+]([O-])=O)C(Cl)=O

InChI

1S/C7H3ClN2O5/c8-7(11)4-1-5(9(12)13)3-6(2-4)10(14)15/h1-3H

InChI key

NNOHXABAQAGKRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,5-Dinitrobenzoyl chloride may be used as a derivatizing agent in the analysis of biogenic amines in fermented foods using high performance liquid chromatography (HPLC).[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Bager et al.
Acta oncologica (Stockholm, Sweden), 54(2), 217-223 (2014-11-11)
As carriers of filaggrin gene (FLG) mutations may have a compromised cervical mucosal barrier against human papillomavirus infection, our primary objective was to study their risk of cervical cancer. We genotyped 586 cervical cancer patients for the two most common
Tatsuya Higashi et al.
Steroids, 72(13), 865-874 (2007-09-04)
The identification and quantification of tetrahydrocorticosterone isomers (THBs; 3alpha,5alpha-, 3beta,5alpha-, 3alpha,5beta- and 3beta,5beta-THB) in rat brains using liquid chromatography (LC)-mass spectrometry (MS) are described. For the identification, the THBs were converted to the atmospheric pressure chemical ionization (APCI)-active derivatives, i.e.
J Kirschbaum et al.
Journal of chromatography. A, 881(1-2), 517-530 (2000-07-25)
The reagent 3,5-dinitrobenzoyl chloride (DNBZ-Cl) was tested for pre-column derivatization of biogenic amines (BAs). Samples were derivatized within 3 min in 1 M NaOH at ambient temperature by adding 2-propanole and 50 mM DNBZ-Cl in acetonitrile. The reaction was terminated
Chemical derivatization to enhance ionization of anabolic steroids in LC-MS for doping-control analysis.
Athanasiadou, I., et al.
Trends in Analytical Chemistry, 42, 137-156 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service