Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72491

Sigma-Aldrich

Ninhydrin

≥95.0% (UV)

Synonym(s):

1,2,3-Indantrione monohydrate, 2,2-Dihydroxy-1,3-indanedione, Trioxohydrindene monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H6O4
CAS Number:
Molecular Weight:
178.14
Beilstein/REAXYS Number:
1910963
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥95.0% (UV)

form

crystals

mp

250 °C (dec.) (lit.)

SMILES string

OC1(O)C(=O)c2ccccc2C1=O

InChI

1S/C9H6O4/c10-7-5-3-1-2-4-6(5)8(11)9(7,12)13/h1-4,12-13H

InChI key

FEMOMIGRRWSMCU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ninhydrin, a tricyclic 1,2,3-trione,[1] is a useful organic building block.[2]

Application

Ninhydrin (Triketohydrindene hydrate) may be used as a reagent for the quantification of amino acids and peptides by photometric method[3] and colorimetric method.[4] It may be used to prepare the ninhydrin reagent, by mixing ninhydrin and hydrindantin (reduced form of ninhydrin) in dimethyl sulfoxide. This reagent is used in manual ninhydrin method for the quantitative determination of amino acids.[5]
Used for the detection of free amino groups in amino acids, peptides and proteins.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photometric ninhydrin method for use in the chromatography of amino acids.
S MOORE et al.
The Journal of biological chemistry, 176(1), 367-388 (1948-10-01)
Isomerization of Ninhydrin-Heterocyclic Ketene Aminal Adducts: Kinetic versus Thermodynamic Control, Solvent Dependency and Mechanism.
Chen N, et al.
European Journal of Organic Chemistry, 28, 6210-6218 (2014)
The determination of amino-acids with ninhydrin.
Yemm EW, et al.
Analyst, 80(948), 209-214 (1955)
Ninhydrin as a building block in scaffold-linked chromophoric dyad construction.
Margetic D, et al.
ARKIVOC (Gainesville, FL, United States), 210, 224-224 (2014)
Yu-Feng Xie et al.
Journal of neurochemistry, 122(3), 605-618 (2012-05-23)
Neurons located in the trigeminal subnucleus caudalis (Vc) play crucial roles in pain and sensorimotor functions in the orofacial region. Because of many anatomical and functional similarities with the spinal dorsal horn (SDH), Vc has been termed the medullary dorsal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service