Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71935

Supelco

Sodium pyrrolidinedithiocarbamate

suitable for suitability for complexometry, ~80%

Synonym(s):

1-Pyrrolidinedithiocarbonic acid sodium salt, Sodium tetramethylenedithiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8NNaS2
CAS Number:
Molecular Weight:
169.24
Beilstein/REAXYS Number:
4028431
EC Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

assay

~80% (NT)
~80%

Quality Level

form

powder

impurities

15-20% water of crystallization

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

suitability

suitable for suitability for complexometry

SMILES string

[Na]SC(=S)N1CCCC1

InChI

1S/C5H9NS2.Na/c7-5(8)6-3-1-2-4-6;/h1-4H2,(H,7,8);/q;+1/p-1

InChI key

KAPVVZCIKRASLC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Complexing agent for heavy metals used for their extraction from aqueous solution[1][2]; Determination of metals by RP-LC of complexes[3][4][5]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Hojabri et al.
Analytical Chemistry, 59, 54-54 (1987)
W. Langseth
Fresenius Journal of Analytical Chemistry, 325, 267-267 (1986)
B. Magyar et al.
Fortschr. Atomspektr. Spurenanal., 1, 659-659 (1984)
J. Stary
Solvent Extraction of Metal Chelates (1964)
G. Schwedt
Chromatographia, 12, 289-289 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service