71882
mEC Broth with Novobiocin
suitable for microbiology, NutriSelect® Plus
Synonym(s):
EC Broth (modified) with Novobiocin, Meat E. coli Broth with Novobiocin, Modified EC Broth with Novobiocin
About This Item
Recommended Products
form
powder
Quality Level
shelf life
limited shelf life, expiry date on the label
composition
bile salts No. 3, 1.12 g/L
di-potassium hydrogen phosphate, 4 g/L
lactose, 5 g/L
novobiocin, 0.02 g/L
peptone, 20 g/L
potassium dihydrogen phosphate, 1.5 g/L
sodium chloride, 5 g/L
manufacturer/tradename
NutriSelect® Plus
technique(s)
microbiological culture: suitable
final pH
6.9±0.2±25 °C
application(s)
environmental
food and beverages
pathogen testing
sample preparation
water monitoring
microbiology
suitability
selective for Escherichia coli
selective for coliforms
selective for enterovirulent E. coli (EEC)
Application
Preparation Note
Footnote
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service