71435
Sodium cyanoborohydride
purum, ≥95.0% (RT)
Synonym(s):
Sodium cyanotrihydridoborate
About This Item
Recommended Products
grade
purum
assay
≥95.0% (RT)
reaction suitability
reagent type: reductant
mp
>242 °C (dec.) (lit.)
solubility
THF: soluble(lit.)
alcohol: soluble(lit.)
hydrocarbons: insoluble(lit.)
water: soluble(lit.)
SMILES string
[Na+].[BH3-]C#N
InChI
1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1
InChI key
CVDUGUOQTVTBJH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Review on sodium cyanoborohydride has been reported.[3][4] It participates in selective reductions with NaCNBH3, modified with zinc chloride,[5] with zinc iodide.[6]
Application
signalword
Danger
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Sol. 1 - Skin Corr. 1B - Water-react 1
supp_hazards
Storage Class
4.3 - Hazardous materials which set free flammable gases upon contact with water
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service