71020
1-Naphthylmethylamine
technical, ≥95.0% (GC)
Synonym(s):
1-(Aminomethyl)naphthalene, 1-Naphthalenemethylamine
About This Item
grade
technical
Quality Level
assay
≥95.0% (GC)
refractive index
n20/D 1.642
n20/D 1.643 (lit.)
bp
290-293 °C (lit.)
density
1.073 g/mL at 25 °C (lit.)
SMILES string
NCc1cccc2ccccc12
InChI
1S/C11H11N/c12-8-10-6-3-5-9-4-1-2-7-11(9)10/h1-7H,8,12H2
InChI key
NVSYANRBXPURRQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service