Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

71014BC

GDS Shiga Toxin Genes (Top 6) Tq

BioControl, Molecular based PCR test for the detection of Shiga Toxin genes in "Top Six" non-O157 Shiga Toxigenic E. coli (STEC) in food

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

shelf life

6‑12 mo.

manufacturer/tradename

BioControl Systems 71014-100

application(s)

food and beverages

compatibility

for use with BioControl GDS

storage temp.

2-8°C

General description

GDS Shiga Toxin Genes (Top 6) Tq is an automated nucleic acid amplification system for the detection of the stx1 and stx2 genes in the following select O-serogroups of E. coli: O103, O111, O121, O145, O26, and O45. GDS utilizes a proprietary IMS-based sample preparation procedure to isolate organisms belonging to these specific 6 O-serogroups prior to genetic analysis for the presence of the stx1 and stx2 genes.

Assurance GDS Shiga Toxin Genes (Top 6) Tq is designed for use in conjunction with Assurance GDS Top 6 STEC (eae) Tq to detect the presence of Shiga Toxigenic E. coli belonging to O serogroups O103, O111, O121, O145, O26, and O45.

Features and Benefits

  • Sequential approach to detecting Top STEC. Following sample preparation with IMS against the Top 6 O groups, samples are analyzed for the presence of (eae) with the GDS Top 6 STEC (eae) assay. Positive samples are subsequently analyzed for Shiga Toxin genes with the GDS Shiga Toxin Genes (Top 6) assay.
  • Single-step 10-18 h enrichment in mEHEC, enrichment media common to all BioControl methods for E.coli O157:H7, Shiga Toxin genes, and Top STEC

Components

  • Shiga Toxin Genes Amplification Tubes Tq
  • Top 6 STEC Concentration Reagent
  • Resuspension Buffer Tq
  • Top STEC Wash Solution

Testing Method

PCR, Immunomagnetic Separation (IMS)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service