Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70725

Supelco

(R)-(−)-1-(1-Naphthyl)ethyl isocyanate

derivatization grade (chiral), LiChropur, ≥99.0%

Synonym(s):

(R)-NEI

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7CH(CH3)NCO
CAS Number:
Molecular Weight:
197.23
Beilstein/REAXYS Number:
4181805
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade (chiral)

Quality Level

assay

≥99.0% (sum of enantiomers, GC)
≥99.0%

form

liquid

optical activity

[α]20/D −47±2°, c = 3.5% in toluene

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

refractive index

n20/D 1.604 (lit.)
n20/D 1.604

bp

106-108 °C/0.16 mmHg (lit.)

density

1.118 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H](N=C=O)c1cccc2ccccc12

InChI

1S/C13H11NO/c1-10(14-9-15)12-8-4-6-11-5-2-3-7-13(11)12/h2-8,10H,1H3/t10-/m1/s1

InChI key

GONOHGQPZFXJOJ-SNVBAGLBSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Chiral derivatizing agent for the indirect resolution of alcohols, thiols and amines by chromatographic separation of diastereomeric derivatives[1][2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

200.1 °F - closed cup

flash_point_c

93.4 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W.H. Pirkle and J. Finn et al.
Asymmetric Synthesis, 1, 87-87 (1983)
D S Dunlop et al.
Analytical biochemistry, 165(1), 38-44 (1987-08-15)
A method for determining the optical purity of amino acids using HPLC and precolumn derivatization is described. (+)-1-(1-Naphthyl)ethyl isocyanate reacts with racemic amino acids, in high yield, to form naphthylethyl carbamoyl derivatives. The resulting diastereoisomeric pairs were separated on reversed-phase
W D Hooper et al.
Journal of chromatography. B, Biomedical applications, 672(1), 89-96 (1995-10-06)
A procedure for the concurrent determination of the (+)- and (-)-enantiomers of sotalol in plasma using high-performance liquid chromatography of diastereomeric derivatives is described. Sotalol is extracted from a 0.5-ml aliquot of plasma at pH 9.3 using ethyl acetate. Atenolol
F J Belas et al.
Biomedical chromatography : BMC, 9(3), 140-145 (1995-05-01)
A high-performance liquid chromatographic assay is described for the separation and quantification of nadolol isomers in human plasma. The isomers were quantified using reverse-phase HPLC and fluorometric detection after derivatization with the chiral reagent R(-)-1-(naphthyl)ethylisocyanate [R(-)-NEI]. The N-isopropyl analogue (one
R A Carr et al.
Journal of chromatography, 566(1), 155-162 (1991-05-03)
A sensitive high-performance liquid chromatographic assay was developed for the determination of tocainide enantiomers in plasma. Following extraction of tocainide from plasma, the enantiomers were derivatized with S-(+)-1-(1-naphthyl)ethylisocyanate. The resulting diastereomers were separated and quantified using normal-phase chromatography with fluorescence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service