70710
(R)-(+)-1-(1-Naphthyl)ethylamine
for chiral derivatization, LiChropur™, ≥99.5%
Synonym(s):
(R)-(+)-α-Methyl-1-naphthalenemethylamine
About This Item
Recommended Products
grade
derivatization grade ((chiral))
for chiral derivatization
product line
ChiraSelect™
assay
≥99.5% (sum of enantiomers, GC)
≥99.5%
form
liquid
optical activity
[α]/D +57±2°, c = 2 in ethanol
optical purity
enantiomeric ratio: ≥99.5:0.5 (GC)
quality
LiChropur™
refractive index
n20/D 1.623 (lit.)
n20/D 1.624
bp
153 °C/11 mmHg (lit.)
density
1.067 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
C[C@@H](N)c1cccc2ccccc12
InChI
1S/C12H13N/c1-9(13)11-8-4-6-10-5-2-3-7-12(10)11/h2-9H,13H2,1H3/t9-/m1/s1
InChI key
RTCUCQWIICFPOD-SECBINFHSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Other Notes
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service