Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70585

Sigma-Aldrich

Benzethonium chloride

tested according to USP

Synonym(s):

Benzethonii chloridum, (Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, Phemerol chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42ClNO2
CAS Number:
Molecular Weight:
448.08
Beilstein/REAXYS Number:
3898548
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

agency

USP/NF
tested according to USP

Quality Level

description

cationic

assay

97.0-103.0% dry basis

form

powder

mol wt

448.08 g/mol

mp

162-164 °C (lit.)

SMILES string

[Cl-].CC(C)(C)CC(C)(C)c1ccc(OCCOCC[N+](C)(C)Cc2ccccc2)cc1

InChI

1S/C27H42NO2.ClH/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H/q+1;/p-1

InChI key

UREZNYTWGJKWBI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 2

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K M Coates et al.
British journal of pharmacology, 134(4), 871-879 (2001-10-19)
1. Ketamine is a dissociative anaesthetic that is formulated as Ketalar, which contains the preservative benzethonium chloride (BCl). We have studied the effects of pure racemic ketamine, the preservative BCl and the Ketalar mixture on human neuronal nicotinic acetylcholine receptors
G Takeoka et al.
Journal of agricultural and food chemistry, 49(7), 3316-3320 (2001-07-17)
Commercial grapefruit seed extracts (GSE) were extracted with chloroform. The solvent was evaporated, and the resulting solid was subsequently analyzed by high-performance liquid chromatography, electrospray ionization mass spectrometry, nuclear magnetic resonance (NMR) spectroscopy, and elemental analysis (by proton-induced X-ray emission
R W Luxton et al.
Clinical chemistry, 35(8), 1731-1734 (1989-08-01)
By using a benzethonium chloride concentration 12-fold that described originally (Clin Chem 1979;25:1317-9), we developed a reliable method suitable for routine measurement of total protein in cerebrospinal fluid. Only 10 microL of sample is required. Reactivity to immunoglobulin G (IgG)
Jacob Pitaro et al.
The Laryngoscope, 123(10), 2521-2525 (2013-08-07)
To determine the ototoxic potential of aluminum acetate (0.5%)/benzethonium chloride (0.03%) otic solution in the chinchilla animal model. A randomized, prospective, controlled study was conducted in the chinchilla animal model. Fourteen female chinchillas were used. After an incision was made
Naoto Kojima et al.
Bioorganic & medicinal chemistry letters, 18(24), 6451-6453 (2008-11-11)
C4-Fluorinated analogues of solamin, an antitumor acetogenin, were synthesized and investigated for their antitumor activities against 39 tumor cell lines. C4-Fluorinated solamins showed more potent growth inhibitory activity against cancer cell lines than solamin.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service