Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

70403

Millipore

AQUANAL-Spectro 3 Photometer

Synonym(s):

Spectro 3 Photometer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

application(s)

environmental

General description

The microprocessor-controlled AQUANAL Spectro 3 Photometer offers complete solutions for the examination of your water and wastewater. It can be used for AQUANAL-plus as well as for all AQUANAL-professional and AQUANAL-professional Vario H reagents. This unit disposes of a substantial user guidance to simplify its handling to an utmost extent.
It operates with the latest LED (Light Emitting Diode) technology. Its measuring range reaches from 380 up to 810 nm with nine different diodes. An excellent reproducibility and a small slit width guarantee precise measuring results. Light emitting diodes emit a stable light and consume only little energy, so that this unit can also be used for a longer period of time. Its advanced optical design and its additional interference filter improving its precision have been developed especially for the AQUANAL Spectro 3 Photometer. The unit is equipped with a large graphic LCD display with a resolution of 128 x 64 pixels.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service