Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

70108

Supelco

AQUANAL-professional Total Nitrogen tube test

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

measuring range

0.5-14 mg/L

application(s)

environmental
general analytical

Packaging

24 reaction tubes, 1 blank tube

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Ox. Liq. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

5.1B - Oxidizing hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service